Thymol 98,5, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 4-Isopropyl-3-hydroxytoluene, 2-Isopropyl-5-methylphenol, 6-Isopropyl-m-cresol

T0501-100G T0501-500G
T0501-100GEA 46.4 GBP
T0501-100G T0501-500G
Thymol 98,5, Sigma-Aldrich®
Thymol

Thymol is generally added in the Hematoxylin and Eosin staining solution to prevent mould growth. It is also suitable for use in antimicrobial screening by the disk diffusion method. It has been used as a test compound in antifungal assay to assess the susceptibility of various fungal isolates.

Formula: 2-[(CH₃)₂CH]C₆H₃-5-(CH₃)OH
MW: 150.22 g/mol
Boiling Pt: 232 °C (lit.)
Melting Pt: 48…51 °C (lit.)
Density: 0.965 g/cm³ (25 °C) (lit.)
Flash Pt: 116 °C (closed cup)
Storage Temperature: Room Temperature
MDL Number: MFCD00002309
CAS Number: 89-83-8
EINECS: 201-944-8
UN: 2430
ADR: 8 III
Merck Index: 13,09476
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b6751e35ab070c8998c9c9e90e15a8b10ccdb9e&vid=3b6751e35ab070c8998c9c9e90e15a8b10ccdb9e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance (Colour) Colorless to White
Appearance (Form) Powder or Crystals
Solubility (Colour) Colorless
Solubility (Turbidity)
50 mg/ml, EtOH
Clear
1H NMR Spectrum Conforms to Structure
Purity (HPLC) ≥98.5 %
Recommended Retest Period 3 years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR