Silvers skin graft knives
Supplier: SWANN MORTON
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
9911 9912
SWAN9911EA 2690 GBP
SWAN9911 SWAN9912
Silvers skin graft knives
Scalpels and Scalpel Assemblies Scalpels
These miniature skin graft handles are to be used with surgical stainless double edged razor blades. Ideal for harvesting small intricate grafts from areas which would not be accessible with the larger standard knives such as the ear, dorsa of the hands or scalp.
  • Non sterile
  • Made from stainless steel
  • Available with solid stainless steel or distinctive blue autoclavable handle
Although used mainly within plastic, reconstructive and oral surgery the graft handles can also be used for the debridement of wounds.
Packaging: Individually boxed.