Scalpel blades
Supplier: SWANN MORTON
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0101 0103 0107 0108 0110 0201 0202 0203 0205 0206 0207 0208 0210 0211 0214 0216 0217 1112 1113 1115 1191 1101 1102 1103 1104 1105 1106 1107 1108 1110 1111
233-5332EA 71 GBP
233-5332 233-5334 233-5345 233-5346 233-5348 233-0023 233-0101 233-0024 233-0025 233-0026 233-0027 233-0028 233-0029 233-0030 233-0031 233-0032 233-0033 SWAN1112 SWAN1113 SWAN1115 SWAN1191 SWAN1101 SWAN1102 SWAN1103 SWAN1104 SWAN1105 SWAN1106 SWAN1107 SWAN1108 SWAN1110 SWAN1111
Scalpel blades
Scalpels and Scalpel Assemblies Scalpel Blades
Carbon steel.
  • Available in sterile individual foil packs or non sterile foil packs of five
  • Quick and easy to assemble
Order Now
SPECIFICATIONS
Environmentally Preferable