SureSwab Rapid Multiple Drug Test Kit
Supplier: SURESCREEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SURESWABRAPID6 SWABRAPID6
SURESURESWABRAPID6EA 400 GBP
SURESURESWABRAPID6 SURESWABRAPID6
SureSwab Rapid Multiple Drug Test Kit
Clinical Diagnostic Supplies Clinical Reagents and Kits
SureSwab rapid kit is quick and easy form of oral drug screening.
  • Optimising collection time
  • Smaller sponge for easier saturation
  • Indicator to display sponge is fully saturated
  • Detection of both parent and metabolic cannabis
  • Slimline design for easier transportation
  • Test up to six parameters simultaneously
The SureSwab Rapid is an easy-to-use screening test to detect multiple drug parameters in oral fluid. It is designed to be less invasive for improved donor experience and enables rapid collection time. A simple solution suitable for workplace, rehab, and clinical testing.
Delivery information: Supplied with test strip, swab collector, procedure card and product IFU.
Caution: For professional use only.