Cole-Parmer® SH-200 Orbital Shakers
Supplier: Cole Parmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
51901-00
STUA51901-00EA 1629.29 GBP
STUA51901-00
Cole-Parmer® SH-200 Orbital Shakers
Shakers Orbital Shakers
Maintain consistent shaking speeds, even over long periods of time.
  • Uniform circular motion and smooth orbital shaking action with 16 mm orbit
  • Built-in timer and digital speed LED display provide reproducible results
  • Can be used in incubators
They are ideal for larger samples and multiwell plates. Easily set and view the speed and the timer on the LED display. Control speed from 30 to 300 min⁻¹. The microprocessor maintains speed accuracy even over long runs. Built-in timer can be set from 1 to 999 minutes. When timer reaches zero, the shakerMore Product Information