Cole-Parmer® Stuart DR-200 Rotating Shaker System Drives
Supplier: Cole Parmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
51600-06 51600-92 51600-93 51600-91 51600-94 51600-95 51600-96 51600-97 51600-98 51600-99
STUA51600-06EA 1257.8 GBP
STUA51600-06 STUA51600-92 STUA51600-93 STUA51600-91 STUA51600-94 STUA51600-95 STUA51600-96 STUA51600-97 STUA51600-98 STUA51600-99
Cole-Parmer® Stuart DR-200 Rotating Shaker System Drives
Shakers Rotating/Rocking Shakers
Versatile design easily switches out mixing drum for different action.
  • Built-in timer
  • Variable speed control: 6 to 60 min⁻¹
  • Change the rotator's capacity with interchangeable drums
  • Clip plates hold various size tubes
The rotating shaker system features a variable-speed drive that accepts one of five drums, providing different mixing actions to a variety of vessels. Set the analog timer from 10 to 60 minutes or operate the unit continuously.