Cole-Parmer® Stuart INC-200 Stuart Hybridisation Incubator/Shakers
Supplier: Cole Parmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
39454-05 39454-95 39454-96
STUA39454-05EA 3695.82 GBP
STUA39454-05 STUA39454-95 STUA39454-96
Cole-Parmer® Stuart INC-200 Stuart Hybridisation Incubator/Shakers
Ovens Hybridisation Ovens
Versatile - features both rotisserie and rocking motions.
  • Forced air circulation combined with microprocessor control result in excellent temperature uniformity
  • Up-and-over door for easy access to the chamber
Use a hybridisation incubator when it is vital to maintain the integrity of the sample while heating it to the exact temperature. The incubator is ideal for nucleic acid hybridisation applications such as Northern (RNA) blots, Southern (DNA) blots, and Western (protein) blots. This compact incubator comes with two built-in shakers. RotisserieMore Product Information