Stuart® Rotary Evaporators
Supplier: Cole Parmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
16340-62 16340-57 16340-58 16340-60
STUA16340-62EA 3299.56 GBP
STUA16340-62 STUA16340-57 STUA16340-58 STUA16340-60
Stuart® Rotary Evaporators
Evaporators Rotary Evaporators
Quality, efficient evaporators at an affordable price.
  • Digital control for reproducible results
  • Versatile - can be orientated for left or right handed users
  • Simple, counterbalanced lift mechanism
  • PTFE/glass liquid pathway for chemical inertness
  • Long-life graphite impregnated PTFE vacuum seal
  • Efficient flask and vapour tube ejection system
The Stuart evaporator range offers simple control and three glassware configurations - diagonal, vertical, or cold finger condensing depending on your application and space requirements.
Choose the diagonalMore Product Information