Cole-Parmer® Stuart MP-200 Digital Melting Point Apparatus
Supplier: Cole Parmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
03011-47 03011-41
STUA03011-47EA 1238.14 GBP
STUA03011-47 STUA03011-41
Cole-Parmer® Stuart MP-200 Digital Melting Point Apparatus
Melting Point Apparatus
No more reading glass thermometers.
  • Select, measure, and display the temperature digitally, ensuring accuracy and repeatability
  • Easily view the melt via magnifying lens with built-in illumination
  • Be more productive - test two samples simultaneously
  • Save time by setting a plateau temperature for initial warm-up
Designed for maximum safety and ease of use. Select, measure, and display the temperature digitally, ensuring accuracy and avoiding the need for a glass thermometer. Test two samples simultaneously, viewing the melt via a magnifying lens withMore Product Information