Stainless steel bubblers, vertical, electropolished with fill-port
Supplier: Strem Chemicals, Inc.
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
96-4998.1
STRE96-4998.1EA 0 GBP
STRE96-4998.1
Stainless steel bubblers, vertical, electropolished with fill-port
Bubblers
Chemical vapor deposition/atomic layer deposition (CVD/ALD) equipment.
  • PCTFE valve stem tip (121 °C)
  • Standard temp. valve
Volatile precursors for CVD.
Ordering information: For dimensions or more information, contact your Avantor representative.