Osteoarthritic human chondrocytes P0 (50-70y), CELLvo™
Supplier: STEMBIOSYS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
OA-100M-000 OA-100M-6WP OA-100M-T75 OA-100M-T150 OA-100F-000 OA-100F-6WP OA-100F-T75 OA-100F-T150
STBSOA-100M-000EA 0 GBP
STBSOA-100M-000 STBSOA-100M-6WP STBSOA-100M-T75 STBSOA-100M-T150 STBSOA-100F-000 STBSOA-100F-6WP STBSOA-100F-T75 STBSOA-100F-T150
Osteoarthritic human chondrocytes P0 (50-70y), CELLvo™
Cells
CELLvo™ Ostoeoarthritic Human Chondrocytes (50-70y) are primary, uncultured cells isolated from recently-diseased cadaver cartilage from donors between 50 and 70 years old. Osteoarthritis status is determined by macroscopic inspection using a modified Outerbridge (1960) scale. Cells are isolated by enzymatic digestion prior to cryopreservation.