Setaflash 3e ActiveCool Flashpoint Tester
Supplier: STANHOPE SETA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
33270-0
STAN33270-0EA 0 GBP
STAN33270-0
Setaflash 3e ActiveCool Flashpoint Tester
Flashpoint Testers
Setaflash Series 3e ActiveCool provides fast and reliable flash point results and requires minimum operator expertise. It has been designed to rapidly and efficiently carry out Flash / No Flash tests to determine the flash point of liquids and semi-solids in the 10 to 135 °C temperature range.
  • Flash/No flash and ramp modes
  • Electric ignitor
  • Flash point test in less than 2 minutes
  • Automatic flash detection and portable operation
  • Electronic peltier cooling
  • Inbuilt touch screen and USB port
  • °C or °F display
The Series 3 ActiveCool Flash Point Tester isMore Product Information