Distillation flask, STANHOPE-SETA
Supplier: STANHOPE SETA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
11962-0 11910-001
STAN11962-0EA 0 GBP
STAN11962-0 STAN11910-001
Distillation flask, STANHOPE-SETA
Flasks Distillation Flasks
For the distillation of petroleum products.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Complies with ASTM D850.