Certified colour reference standards for spectrophotometers
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
9423UV95520E 9423UV95100E 401322989841
SPET9423UV95520EEA 2260 GBP
SPET9423UV95520E SPET9423UV95100E SPET401322989841
Certified colour reference standards for spectrophotometers
Standards Colour Reference and Spectroscopy Standards
These colour reference standards can be used with UV-Vis spectroscopy.
  • Each standard includes a calibration certificate
Certifications: Traceable to ISO/IEC Standard 17025 accredited calibration process.