Gum tragacanth, powder NF

Supplier: Spectrum Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Tragacanth, Tragacanth gum

TR105-12KG TR105-125GM TR105-2.5KG TR105-500GM TR105-50KGBL
SPCMTR105-12KGEA 5530 GBP
SPCMTR105-12KG SPCMTR105-125GM SPCMTR105-2.5KG SPCMTR105-500GM SPCMTR105-50KGBL
Gum tragacanth, powder NF
Gum tragacanth

Tragacanth, Powder, NF is used as an emulsifier, thickener, stabiliser, and texturant additive.


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).

Storage Temperature: Ambient
MDL Number: MFCD00131255
CAS Number: 9000-65-1
EINECS: 232-552-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR