(±)-1,2-Propanediol 99,5 USP

Supplier: Spectrum Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Propylene glycol, 1,2-Propanediol, (±)-1,2-Propylene glycol, 1,2-Dihydroxypropane, (±)-1,2-Dihydroxy propane

PR130-20LTPL PR130-4LTGL PR130-4LTPL PR130-200LTBL PR130-500MLPL
SPCMPR130-20LTPLEA 1280 GBP
SPCMPR130-20LTPL SPCMPR130-4LTGL SPCMPR130-4LTPL SPCMPR130-200LTBL SPCMPR130-500MLPL
(±)-1,2-Propanediol 99,5 USP
Propylene glycol

Propylene Glycol, USP is used in pharmaceutical products as an excipient solvent in oral and topical drug products. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: CH₃CH(OH)CH₂OH
MW: 76.1 g/mol
Boiling Pt: 187.6
Melting Pt: -59
Flash Pt: 99
Storage Temperature: Ambient
MDL Number: MFCD00064272
CAS Number: 57-55-6
Merck Index: 13,07947
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2af195b87767feb8d1baca34925153d6f97ae2d&vid=a598a9042847dc3ce52daed1a9326b592b163170&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.5 % min.
Specific Gravity @ 25°C 1.035 - 1.037
Acidity To Pass Test
Water Determination max. 0.2 %
Residue On Ignition max. 0.007 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR