L(+)-Potassium hydrogen tartrate USP

Supplier: Spectrum Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Potassium hydrogen L-tartrate, Potassium acid tartrate, Potassium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate, Potassium Bitartrate, (R,R)-(+)-Tartaric acid potassium salt, L(+)-Tartaric acid potassium salt

P1801-12KG P1801-2.5KG P1801-50KGBL
SPCMP1801-12KGEA 1450 GBP
SPCMP1801-12KG SPCMP1801-2.5KG SPCMP1801-50KGBL
L(+)-Potassium hydrogen tartrate USP
L(+)-Potassium hydrogen tartrate
Formula: KOOC(CHOH)₂COOH
Melting Pt: 250 °C
Density: 1.943 g/cm³ (20 °C)
MDL Number: MFCD00065392
CAS Number: 868-14-4
EINECS: 212-769-1
Merck Index: 13,07697
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=355f72f319f5589f12d60c6dfffecdec31cee083&vid=4a750ad5d4a91fd4c2885832ee80b7b64cdcc42a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR