Polyvinylpyrrolidone, M.W. 1 300 000 USP (K90)

Supplier: Spectrum Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Povidone, Polyvinylpyrrolidon, Polyvidone, PVP

P1416-12KG P1416-2.5KG P1416-50KGBL
SPCMP1416-12KGEA 5330 GBP
SPCMP1416-12KG SPCMP1416-2.5KG SPCMP1416-50KGBL
Polyvinylpyrrolidone, M.W. 1 300 000 USP (K90)
Polyvinylpyrrolidone

Melamine statement (available upon request)
BSE/TSE statement (available upon request)

Formula: (C₆H₉NO)n
Storage Temperature: Ambient
MDL Number: MFCD01076626
CAS Number: 9003-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR