Potassium iodide ≥99.0%, granular FCC

Supplier: Spectrum Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

P1344-2.5KG P1344-500GM
SPCMP1344-2.5KGEA 3460 GBP
SPCMP1344-2.5KG SPCMP1344-500GM
Potassium iodide ≥99.0%, granular FCC
Potassium iodide

Potassium Iodide, Granular, FCC is used as a source for dietary iodine. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark;Health hazard

Formula: Kl
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0

Order Now

Specification Test Results

Assay (Dried Basis) 99.0 - 101.5%
Iodate (IO₃) max. 4 mg/kg
Lead (Pb) max. 4 mg/kg
Loss On Drying max. 1 %
Nitrate, Nitrite, and Ammonia To Pass Test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR