Methyl-4-hydroxybenzoate ≥98.0% NF

Supplier: Spectrum Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ME163-500GM ME163-50KGBL
SPCMME163-500GMEA 197 GBP
SPCMME163-500GM SPCMME163-50KGBL
Methyl-4-hydroxybenzoate ≥98.0% NF
Methyl 4-hydroxybenzoate

Methylparaben, NF is an excipient used as a preservative. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark.

Formula: HOC₆H₄CO₂CH₃
Storage Temperature: Ambient
MDL Number: MFCD00002352
CAS Number: 99-76-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84321052052f4be2a53d8d7ebac7fabe6fc5a435&vid=ede765fad275b2515a8c57ba4499bff313a18983&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (C₈H₈O₃) 98.0 - 102.0 %
Color of Solution To Pass Test
Acidity To Pass Test
Residue On Ignition max. 0.1 %
Related Substances To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR