Isopropyl myristate NF

Supplier: Spectrum Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Tetradecanoic acid isopropyl ester, Myristic acid isopropyl ester, Isopropyl Myristate, Isopropyl tetradecanoate, Tetradecylic acid isopropyl ester

IS145-200LTBL IS145-20LTMT IS145-4LTGL
SPCMIS145-200LTBLEA 5390 GBP
SPCMIS145-200LTBL SPCMIS145-20LTMT SPCMIS145-4LTGL
Isopropyl myristate NF
Isopropyl myristate
Formula: C₁₇H₃₄O₂
Boiling Pt: 320 °C (1013 hPa)
Melting Pt: 0…1 °C
Density: 0.852 g/cm³ (20 °C)
Flash Pt: 150 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008982
CAS Number: 110-27-0
EINECS: 203-751-4
Merck Index: 13,05235

Order Now

Specification Test Results

Assay 90% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR