Erythrosin B disodium salt

Supplier: Spectrum Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Erythrosine sodium, C.I. 45430, Erythrosin B, Sodium 2',4',5',7'-tetraiodo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate), Erythrosine 3B, FD&C Red #3, Erythrosin 239, Acid Red 51

FD135-100GM FD135-25GM FD135-500GM
SPCMFD135-100GMEA 209 GBP
SPCMFD135-100GM SPCMFD135-25GM SPCMFD135-500GM
Erythrosin B disodium salt
Erythrosin B disodium salt

FD and C Red No. 3 | Non-graded FD and C Red No. 3 supplied by Spectrum is indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂₀H₆I₄Na₂O₅
MW: 879.86 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00144257
CAS Number: 16423-68-0
EINECS: 240-474-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR