Oil of castor USP

Supplier: Spectrum Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

CA208-200LTBL CA208-20LTMT CA208-4LTGL CA208-500MLGL
SPCMCA208-200LTBLEA 5290 GBP
SPCMCA208-200LTBL SPCMCA208-20LTMT SPCMCA208-4LTGL SPCMCA208-500MLGL
Oil of castor USP
Oil of castor

BSE/TSE statement (available upon request)
Allergen statement (available upon request)

Boiling Pt: 313 °C (1013 hPa)
Melting Pt: –10 °C
Density: 0.95 g/cm³ (20 °C)
Flash Pt: >113 °C (closed cup)
MDL Number: MFCD00130746
CAS Number: 8001-79-4
EINECS: 232-293-8
Merck Index: 13,01908

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR