Calcium sulphate dihydrate NF

Supplier: Spectrum Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

CA170-12KG CA170-2.5KG CA170-45KGBL
SPCMCA170-12KGEA 1090 GBP
SPCMCA170-12KG SPCMCA170-2.5KG SPCMCA170-45KGBL
Calcium sulphate dihydrate NF
Calcium sulphate dihydrate
Formula: CaSO₄·2H₂O
Density: 2.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149625
CAS Number: 10101-41-4
EINECS: 231-900-3
Merck Index: 13,01711

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR