Calcium chloride dihydrate ≥99% USP, Ph. Eur., BP, JP

Supplier: Spectrum Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

CA140-500GM
SPCMCA140-500GMEA 199 GBP
SPCMCA140-500GM
Calcium chloride dihydrate ≥99% USP, Ph. Eur., BP, JP
Calcium chloride dihydrate

Calcium Chloride, Dihydrate, USP, EP, BP, JP is used in treatment of hypocalcemia, electrolyte depletion, and hyperkalemia. All Spectrum Chemical USP, EP, BP, JP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: CaCl₂·2H₂O
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 176 °C
Density: 1,85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149613
CAS Number: 10035-04-8
EINECS: 233-140-8
Merck Index: 13,01660

Order Now

Specification Test Results

Assay 99% Min.

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR