Calcium citrate tetrahydrate, powder USP

Supplier: Spectrum Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: tri-Calcium dicitrate tetrahydrate, Calcium citrate tribasic tetrahydrate

C1102-125LBBL C1102-12KG C1102-2.5KG
SPCMC1102-125LBBLEA 3510 GBP
SPCMC1102-125LBBL SPCMC1102-12KG SPCMC1102-2.5KG
Calcium citrate tetrahydrate, powder USP
Calcium citrate tetrahydrate

Form: Powder

Formula: C₁₂H₁₀Ca₃O₁₄·4H₂O
MDL Number: MFCD00150786
CAS Number: 5785-44-4
EINECS: 212-391-7

Order Now

Specification Test Results

Assay 97.5% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR