Cleanroom Wipes, MicroPure
Supplier: SONTARA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1000003957_U 1000003838_U 1000001898 1000003980 1000001912
SONT1000003957_UEA 557 GBP
SONT1000003957_U SONT1000003838_U SONT1000001898 SONT1000003980 SONT1000001912
Cleanroom Wipes, MicroPure
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Sontara® Cleanroom wipes are specially designed for the challenging task of cleaning a cleanroom - ideal for electronic manufacturing, pharmaceutical, biotechnology applications and surface mount technology. These micro-level cleaning fabrics are 'virtually' lint-free and naturally absorbent, without binders or other additives.
  • Exceptionally clean wipe family with extremely low particle generation, low extractables and ion levels
  • Made with no binders, chemicals or adhesives
  • Special pattern for excellent pick-up
  • Excellent wet and dry strength
  • Solvent
More Product Information