TriLink® 2'-O-Methyl-N6-Methyladenosine-5'-Triphosphate

Supplier: TriLink BioTechnologies
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

N-1112-1 N-1112-10 N-1112-100 N-1112-5
SOLUN-1112-1EA 68.6 GBP
SOLUN-1112-1 SOLUN-1112-10 SOLUN-1112-100 SOLUN-1112-5
TriLink® 2'-O-Methyl-N6-Methyladenosine-5'-Triphosphate
Nucleic Acid Reagents Nucleotides

  • Grade: RUO


Caution: Item will be delivered at or below –20 °C and should be stored at or below –20 °C.

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=923da848c5aa1771fd575a9a582866655a3e4180&vid=923da848c5aa1771fd575a9a582866655a3e4180&prid=web&clid=SAPDAM
New Product

Order Now

SPECIFICATIONS

Environmentally Preferable
Application

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR