TriLink® 2'-O-Methylcytidine-5'-Triphosphate
Supplier: TriLink BioTechnologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
N-1016-1 N-1016-10 N-1016-5
SOLUN-1016-1EA 19.2 GBP
SOLUN-1016-1 SOLUN-1016-10 SOLUN-1016-5
TriLink® 2'-O-Methylcytidine-5'-Triphosphate
Nucleic Acid Reagents Nucleotides
  • Grade: RUO
Caution: Item will be delivered at or below –20 °C and should be stored at or below –20 °C.