TriLink® N6-Methyladenosine-5'-Triphosphate

Supplier: TriLink BioTechnologies
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

N-1013-1 N-1013-10 N-1013-5
SOLUN-1013-1EA 85.9 GBP
SOLUN-1013-1 SOLUN-1013-10 SOLUN-1013-5
TriLink® N6-Methyladenosine-5'-Triphosphate
Nucleic Acid Reagents Nucleotides

N6-methyl adenosine (N6-methyl ATP) is a base modified analog of adenosine and is found as a minor nucleoside in natural RNAs (Meyer et al.). N6-methyl ATP can substitute for ATP in some biological systems, and is a potent agonist for P2Y-purinoceptors in the guinea pig, taenia coli (Burnstock et al.).


  • Grade: RUO


Caution: Item will be delivered at or below –20 °C and should be stored at or below –20 °C.

New Product

Order Now

SPECIFICATIONS

Environmentally Preferable
Application

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR