TriLink® CleanCap® mCherry mRNA (5 moU)

Supplier: TriLink BioTechnologies
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

L-7203-100 L-7203-1000 L-7203-5
SOLUL-7203-100EA 293 GBP
SOLUL-7203-100 SOLUL-7203-1000 SOLUL-7203-5
TriLink® CleanCap® mCherry mRNA (5 moU)
Protein Expression Protein Expression Reagents

mCherry mRNA encodes the fluorescent protein, mCherry, which is derived from DsRed, a protein found in Discosoma sp. mCherry is a monomeric fluorophore with a peak absorption at 587 nm and emission at 610 nm. It is stable and resistant to photobleaching.


  • Capped with CleanCap® AG cap analog, modified with 5-methoxyuridines

  • Cap 1 structure with >95% capping efficiency for superior in vivo activity compared to Cap-0 mRNAs

  • Manufactured with TriLink’s proprietary CleanScript® method to lower dsRNA for improved performance

  • Ready to use in downstream mRNA applications and formulations

  • Grade: RUO


This mRNA is capped using CleanCap® Reagent AG, TriLink's co-transciptional capping technology, which results in the naturally occurring Cap-1 structure with >95% capping efficiency. It is manufactured using TriLink’s CleanScript® method, polyadenylated, modified with 5-methoxyuridine, and optimised for mammalian systems. It mimics a fully processed mature mRNA.


CleanCap Reagent AG produces a base-modified Cap-1 mRNA, which shows superior in vivo activity compared to Cap-0 mRNA produced by legacy capping methods such as mCap or anti-reverse cap analog (ARCA).


5-methoxyuridine (5moU) is a modified uridine that can reduce immunogenic response and enhance translational efficiency of mRNAs. These properties can result in safer mRNA and increased protein expression.


CleanScript method is TriLink’s proprietary in vitro transcription process that is optimised to minimise dsRNA formation and increase mRNA activity in vivo.


Caution: Item will be delivered at or below −40 °C and should be stored at or below −40 °C.

New Product

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR