Reagent reservoirs for single and multi channel pipettes
Supplier: Socorex Isba
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
330.01 330.12.9 330.01L 330.08.9 330.08 330.08XL 330.12 330.12XL
SOCO330.01EA 29.2 GBP
SOCO330.01 613-0549 SOCO330.01L SOCO330.08.9 SOCO330.08 SOCO330.08XL SOCO330.12 SOCO330.12XL
Reagent reservoirs for single and multi channel pipettes
Reagent Reservoirs
These reagent reservoirs are either made from PP or from PS.
V-shape reservoirs (PP), 75 m:
- Reusable and autoclavable PP
- Embossed graduations guarantee long lasting legibility
- Round corners enable convenient reagent pouring
- Bottom shape ensures bench stability
- Maximal flexibility for routine work with single and multi-channel pipettes
V-shape reservoirs (PP), with lid, 125 ml:
- Reusable and autoclavable PP
- Hinged lid closure
- Microplate format guarantees excellent stability
- Stackable for space saving storage
- Ideal for all types of applications with single and multi-channelMore Product Information