Flocked swabs
Supplier: STERILIN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
501CS01 502CS01 503CS01 511CS01 516C 516CS01 551C 552C 553C 525CS01 520CS01
SLIN501CS01EA 197 GBP
SLIN501CS01 SLIN502CS01 SLIN503CS01 SLIN511CS01 SLIN516C SLIN516CS01 SLIN551C 300-0224 SLIN553C SLIN525CS01 SLIN520CS01
Flocked swabs
Swabs Swabs for Sampling
This innovative nylon fibre technology and unbeatable sample collection and release make the swabs PCR compatible for molecular analysis.
  • Manufactured from completely inert components
  • Moulded breakpoint that is easily breakable
  • Collection vial in various packaging
For forensic purposes product is available certified human DNA free.
Certifications: Certified RNAse- and DNAse-free.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904