Caps for sample tubes, tamper evident
Supplier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
T550W T550WOS T552-7WTP T552WTP T552-30WTP T552-30W T552W
720-0574EA 84.2 GBP
720-0574 216-1007 SIMPT552-7WTP SIMPT552WTP SIMPT552-30WTP SIMPT552-30W SIMPT552W
Caps for sample tubes, tamper evident
Closures Test/Sample Tube Closures
Caps made of PE feature a long skirt and a super fast thread design allowing them to be removed or sealed with a single turn. Caps are designed with flexible sealing lip which provides leakproof seal.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904