Thermo-conductive racks, ChillBlock™
Supplier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
S700-6 S700-50 S700-80 S700-1 S700-10 S700-90 S700-14 S700-16 S700-35 S700-4 S700-56
SIMPS700-6EA 757 GBP
SIMPS700-6 SIMPS700-50 SIMPS700-80 SIMPS700-1 SIMPS700-10 SIMPS700-90 SIMPS700-14 SIMPS700-16 SIMPS700-35 SIMPS700-4 SIMPS700-56
Thermo-conductive racks, ChillBlock™
Racks Tube Racks
ChillBlock™ thermo-conductive metal alloy tube racks eliminate inconsistencies that result from inserting tubes directly into ice, dry ice, alcohol baths, water baths, and other common laboratory temperature sources.
  • Anodised surface that is resistant to rust, corrosion, and abrasion
  • Alphanumeric well identification facilitates tube location
  • Racks can be autoclaved or cleaned with disinfectant
ChillBlock™ tube racks are available in a variety of sizes for vessels such as microcentrifuge tubes, cryogenic vials, PCR tubes, SBS-compliant strips andMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable