Weighing dishes, PS, anti-static, hexagonal
Supplier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
D252-1 D252-2 D252-3 D252-4
SIMPD252-1EA 42.3 GBP
SIMPD252-1 SIMPD252-2 SIMPD252-3 SIMPD252-4
Weighing dishes, PS, anti-static, hexagonal
Boats Weighing Boats
PS, hexagonal
  • Suitable for weighing of static-affected samples
  • Stackable for easy storage
  • Temperature resistant up to +80 °C
Excellent for handling solids or liquids during weighing. Easily bent into pouring spouts, the dishes enable non-spill transfer. Molded hexagonal design provides greater balance protection and safety.