Viscosity cups, Zahn
Supplier: Industrial Physics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
VF2227 VF2230 VF2228 VF2226 VF2229
SIMEVF2227EA 223 GBP
SIMEVF2227 SIMEVF2230 SIMEVF2228 SIMEVF2226 SIMEVF2229
Viscosity cups, Zahn
Viscometers Flow Cup Viscometers
Stainless steel flow cup viscometers with fixed nozzle for measuring coatings and other fluids during application or production.
Certifications: ASTM D 1084, D 4212