Flow cups with fixed nozzle 'Ford'
Supplier: Industrial Physics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
VF2032 VF2030 VF2031 VF2033
SIMEVF2032EA 228 GBP
SIMEVF2032 SIMEVF2030 SIMEVF2031 SIMEVF2033
Flow cups with fixed nozzle 'Ford'
Viscometers Flow Cup Viscometers
Titanium anodised aluminium flow cups for measurement of relative viscosity.
  • Each cup has an engraved unique serial number
  • Design of cup and orifice facilitate cleaning process
  • Deep well surrounding the top of the cup to catch any overflow
Certifications: ASTM D1200
Delivery information: Supplied with plastic case and internal soft protective material.
Order Now
SPECIFICATIONS
Environmentally Preferable