Viscosity cups with nozzle
Supplier: Industrial Physics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
VF1999 VF2003 VF2004 VF2001 VF2000 VF2015
769-1110EA 173 GBP
769-1110 769-1121 769-1125 SIMEVF2001 SIMEVF2000 SIMEVF2015
Viscosity cups with nozzle
Viscometers Flow Cup Viscometers
Anodised aluminium and stainless steel viscometer cups with nozzle for break-thread flow time comparative measurements.
  • Easy cleaning
  • Engraved with a unique serial number
Typically used for the consistency of paintings, varnishes and similar products.
Certifications: DIN 53211
Ordering information: The stand should be ordered separately.