Surface roughness gauge
Supplier: Industrial Physics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SP1562
SIMESP1562EA 487 GBP
SIMESP1562
Surface roughness gauge
Coating Testers
Surface profile roughness gauge for ultrafast measurement of the peaks and valleys of a surface.
The shape of the base is suitable to measure the height of welds.
Certifications: ASTM D4417
Delivery information: Supplied with a reference glass plate, an Allen key and an extra battery in a leather pouch.