trans,trans-2,4-Hexadienal, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ300

W342904-100G W342904-1KG W342904-4KG W342904-SAMPLE
SIALW342904-100GEA 108 GBP
SIALW342904-100G SIALW342904-1KG SIALW342904-4KG SIALW342904-SAMPLE
trans,trans-2,4-Hexadienal, Sigma-Aldrich®
trans,trans-2,4-Hexadienal
Formula: C₆H₈O
MDL Number: MFCD00007004
CAS Number: 142-83-6
EINECS: 205-564-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR