Dimethyl acetal, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Acetaldehyde dimethyl acetal, 1,1-Dimethoxyethane

M-Clarity™ quality level = MQ300

W342602-100G W342602-1KG W342602-SAMPLE
SIALW342602-100GEA 128 GBP
SIALW342602-100G SIALW342602-1KG SIALW342602-SAMPLE
Dimethyl acetal, Sigma-Aldrich®
Dimethyl acetal
Formula: C₄H₁₀O₂
Boiling Pt: 64 °C (1013 hPa)
Melting Pt: –113 °C
Density: 0.8505 g/cm³ (20 °C)
MDL Number: MFCD00008493
CAS Number: 534-15-6
EINECS: 208-589-8
UN: 2377
ADR: 3,II
Merck Index: 13,03253

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR