Pyridine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Azabenzene, Azine

M-Clarity™ quality level = MQ300

W296600-10KG-K W296600-1KG-K W296600-25KG-K
SIALW296600-10KG-KEA 578 GBP
SIALW296600-10KG-K SIALW296600-1KG-K SIALW296600-25KG-K
Pyridine, Sigma-Aldrich®
Pyridine
Formula: C₅H₅N
Boiling Pt: 115 °C (1013 hPa)
Melting Pt: –44 °C
Density: 0.983 g/cm³ (20 °C)
Flash Pt: 17 °C
Storage Temperature: Ambient
MDL Number: MFCD00011732
CAS Number: 110-86-1
EINECS: 203-809-9
UN: 1282
ADR: 3,II
Merck Index: 13,08060
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f9c5934940218027dae0d80a1f98b6d5179c0a8&vid=9f2ef46a26e488d366e82869b3acb96652b5dab1&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR