Propyl butyrate

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ300

W293415-100G-K W293415-1KG-K W293415-25G-K W293415-4KG-K
SIALW293415-100G-KEA 128 GBP
SIALW293415-100G-K SIALW293415-1KG-K SIALW293415-25G-K SIALW293415-4KG-K
Propyl butyrate
Propyl butyrate
Formula: C₇H₁₄O₂
MDL Number: MFCD00009396
CAS Number: 105-66-8
EINECS: 203-320-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR