Polysorbate 20, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Polyoxyethylene-20-Sorbitan Monolaurate, Sorbitan monolaurate, ethoxylated

M-Clarity™ quality level = MQ300

W291501-10KG W291501-1KG W291501-25KG W291501-SAMPLE
SIALW291501-10KGEA 282 GBP
SIALW291501-10KG SIALW291501-1KG SIALW291501-25KG SIALW291501-SAMPLE
Polysorbate 20, Sigma-Aldrich®
Polysorbate 20
Formula: C₅₈H₁₁₄O₂₆
Storage Temperature: Ambient
MDL Number: MFCD00165986
CAS Number: 9005-64-5
EINECS: 500-018-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR