Oleic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: (9Z)-9-Octadecenoic acid

MQ300

W281506-10KG-K W281506-1KG-K W281506-20KG-K W281506-4KG-K
SIALW281506-10KG-KEA 329 GBP
SIALW281506-10KG-K SIALW281506-1KG-K SIALW281506-20KG-K SIALW281506-4KG-K
Oleic acid, Sigma-Aldrich®
Oleic acid
Formula: C₁₈H₃₄O₂
MW: 282.47 g/mol
Boiling Pt: 286
Melting Pt: 16.7
Density: 0.8946 g/cm³ (20 °C)
Flash Pt: 180
Storage Temperature: Freezer
MDL Number: MFCD00064242
CAS Number: 112-80-1
EINECS: 204-007-1
Merck Index: 13,06898
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5aecc5dce4d0318386a45d7adb7c5822631010a&vid=892a2df4ecab3679b06983b84cd27ad334ef961f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR