Methyl butyrate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ300

W269301-1KG-K W269301-20KG-K W269301-5KG-K
SIALW269301-1KG-KEA 104 GBP
SIALW269301-1KG-K SIALW269301-20KG-K SIALW269301-5KG-K
Methyl butyrate, Sigma-Aldrich®
Methyl butyrate
Formula: C₅H₁₀O₂
Boiling Pt: 102 °C (1013 hPa)
Melting Pt: –84 °C
Density: 0.8984 g/cm³ (20 °C)
Flash Pt: 14 °C
MDL Number: MFCD00009391
CAS Number: 623-42-7
EINECS: 210-792-1
UN: 1237
ADR: 3,II
Merck Index: 13,06062

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR