1-Hexadecanol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Cetyl alcohol, Palmityl alcohol

M-Clarity™ quality level = MQ300

W255408-10KG W255408-1KG W255408-SAMPLE
SIALW255408-10KGEA 452 GBP
SIALW255408-10KG SIALW255408-1KG SIALW255408-SAMPLE
1-Hexadecanol, Sigma-Aldrich®
1-Hexadecanol
Formula: H₃C(CH₂)₁₄CH₂OH
Boiling Pt: 305…330 °C (1013 hPa)
Melting Pt: 46…52 °C
Density: 0.81 g/cm³ (20 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004760
CAS Number: 36653-82-4
EINECS: 253-149-0
Merck Index: 13,02037

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR