Heptyl butyrate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ300

W254908-4KG-K W254908-9KG-K
SIALW254908-4KG-KEA 690 GBP
SIALW254908-4KG-K SIALW254908-9KG-K
Heptyl butyrate, Sigma-Aldrich®
Heptyl butyrate
Formula: C₁₁H₂₂O₂
Boiling Pt: 225…226 °C (1013 hPa)
Melting Pt: –58 °C
Density: 0.864 g/cm³ (25 °C)
MDL Number: MFCD00048908
CAS Number: 5870-93-9
EINECS: 227-526-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR