Ethyl-3-phenylglycidate (mixture of cis and trans isomers), Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Ethyl 3-phenylglycidate

M-Clarity™ quality level = MQ300

W245402-SAMPLE W245402-1KG W245402-5KG
SIALW245402-SAMPLEEA 64.2 GBP
SIALW245402-SAMPLE SIALW245402-1KG SIALW245402-5KG
Ethyl-3-phenylglycidate (mixture of cis and trans isomers), Sigma-Aldrich®
Ethyl 3-phenylglycidate (mixture of cis and trans isomers)
Formula: C₁₁H₁₂O₃
MDL Number: MFCD00005123
CAS Number: 121-39-1
EINECS: 204-467-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR